Assay Details
| Gene Symbol: acp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099250.1 | NP_001092720.1 | 170 | 96 | ||
| GenBank mRNA | BC139891.1 | - | 170 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acp7| Gene Symbol: acp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099250.1 | NP_001092720.1 | 170 | 96 | ||
| GenBank mRNA | BC139891.1 | - | 170 | 96 | ||
Set Membership: |
|