Assay Details
| Gene Symbol: adcy2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099987.1 | NP_001093457.1 | 17 - 18 | 2333 | 73 | |
| XM_005170038.4 | XP_005170095.1 | 17 - 18 | 2527 | 73 | ||
| XM_021468083.1 | XP_021323758.1 | 13 - 14 | 1472 | 73 | ||
More Information
Set Membership: |
|

adcy2b
Genomic Map