Assay Details
| Gene Symbol: panx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100030.2 | NP_001093500.1 | 5 - 6 | 1080 | 73 | |
| XM_021476289.1 | XP_021331964.1 | 4 - 5 | 909 | 73 | ||
More Information
Set Membership: |
|

panx1b
Genomic Map