Assay Details
| Gene Symbol: mpp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327790.1 | NP_001314719.1 | 7 - 8 | 955 | 63 | |
| XM_021470936.1 | XP_021326611.1 | 7 - 8 | 837 | 63 | ||
| GenBank mRNA | BC134139.1 | - | 7 - 8 | 955 | 63 | |
| BC152524.1 | - | 4 - 5 | 233 | 63 | ||
More Information
Set Membership: |
|

mpp2a
Genomic Map