Assay Details
| Gene Symbol: mpp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327790.1 | NP_001314719.1 | 12 - 13 | 1623 | 60 | |
| XM_021470936.1 | XP_021326611.1 | 12 - 13 | 1505 | 60 | ||
| GenBank mRNA | BC152524.1 | - | 9 - 10 | 901 | 60 | |
More Information
Set Membership: |
|

mpp2a
Genomic Map