Assay Details
| Gene Symbol: casp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109712.1 | NP_001103182.1 | 5 - 6 | 692 | 138 | |
| GenBank mRNA | BC151948.1 | - | 5 - 6 | 692 | 138 | |
| MG958010.1 | - | 5 - 6 | 552 | 138 | ||
More Information
Set Membership: |
|

casp23
Genomic Map