Assay Details
| Gene Symbol: alcama | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131000.1 | NP_571075.1 | 5 - 6 | 694 | 96 | |
| XM_005172635.4 | XP_005172692.1 | 5 - 6 | 720 | 96 | ||
| GenBank mRNA | BC050482.1 | - | 5 - 6 | 630 | 96 | |
| BC071276.1 | - | 5 - 6 | 695 | 96 | ||
| CU458879.1 | - | 5 - 6 | 535 | 96 | ||
| CU458962.1 | - | 5 - 6 | 641 | 96 | ||
| L25273.1 | - | 5 - 6 | 694 | 96 | ||
More Information
Set Membership: |
|

alcama
Genomic Map