Assay Details
| Gene Symbol: lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131002.2 | NP_571077.2 | 2 - 3 | 547 | 68 | |
| GenBank mRNA | AJ005936.1 | - | 2 - 3 | 541 | 68 | |
| BC162105.1 | - | 2 - 3 | 401 | 68 | ||
More Information
Set Membership: |
|

lmnb2
Genomic Map