Assay Details
| Gene Symbol: spon2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131008.4 | NP_571083.4 | 3 - 4 | 527 | 69 | |
| GenBank mRNA | AB006085.1 | - | 2 - 3 | 1098 | 69 | |
| BC078279.1 | - | 3 - 4 | 532 | 69 | ||
| BC121778.1 | - | 3 - 4 | 527 | 69 | ||
| BC165308.1 | - | 2 - 3 | 474 | 69 | ||
More Information
Set Membership: |
|

spon2b
Genomic Map