Assay Details
| Gene Symbol: spon2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131008.4 | NP_571083.4 | 5 - 6 | 901 | 98 | |
| GenBank mRNA | AB006085.1 | - | 4 - 5 | 1472 | 98 | |
| BC078279.1 | - | 5 - 6 | 906 | 98 | ||
| BC121778.1 | - | 5 - 6 | 901 | 98 | ||
| BC165308.1 | - | 4 - 5 | 848 | 98 | ||
More Information
Set Membership: |
|

spon2b
Genomic Map