Assay Details
| Gene Symbol: fzd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131139.2 | NP_571214.2 | 1 | 1932 | 93 | |
| GenBank mRNA | AF336123.1 | - | 1 | 93 | ||
| BC068322.1 | - | 1 | 1825 | 93 | ||
More Information
Set Membership: |
|

fzd7a
Genomic Map