Assay Details
| Gene Symbol: ran | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131309.3 | NP_571384.1 | 5 - 6 | 614 | 86 | |
| XM_005173094.4 | XP_005173151.1 | 5 - 6 | 601 | 86 | ||
| GenBank mRNA | BC050517.1 | - | 5 - 6 | 86 | ||
| BC058047.1 | - | 4 - 5 | 86 | |||
| EH499718.1 | - | 5 - 6 | 578 | 86 | ||
| U61395.1 | - | 4 - 5 | 86 | |||
More Information
Set Membership: |
|

ran
Genomic Map