Assay Details
| Gene Symbol: mef2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131312.2 | NP_571387.2 | 5 - 6 | 888 | 98 | |
| XM_005169228.4 | XP_005169285.1 | 4 - 5 | 818 | 98 | ||
| XM_005169229.4 | XP_005169286.1 | 5 - 6 | 936 | 98 | ||
| XM_005169232.4 | XP_005169289.1 | 5 - 6 | 932 | 98 | ||
| XM_009305677.3 | XP_009303952.1 | 4 - 5 | 821 | 98 | ||
| XM_009305679.3 | XP_009303954.1 | 3 - 4 | 405 | 98 | ||
| XM_009305680.3 | XP_009303955.1 | 3 - 4 | 401 | 98 | ||
| XM_009305681.3 | XP_009303956.1 | 5 - 6 | 937 | 98 | ||
| GenBank mRNA | BC059188.1 | - | 5 - 6 | 888 | 98 | |
| BC070007.1 | - | 4 - 5 | 755 | 98 | ||
| KF932281.1 | - | 3 - 4 | 408 | 98 | ||
| KF932282.1 | - | 3 - 4 | 408 | 98 | ||
| KT020763.1 | - | 3 - 4 | 514 | 98 | ||
| U66569.1 | - | 3 - 4 | 459 | 98 | ||
More Information
Set Membership: |
|

mef2ca
Genomic Map