Assay Details
| Gene Symbol: mef2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131312.2 | NP_571387.2 | 7 - 8 | 1122 | 123 | |
| XM_005169228.4 | XP_005169285.1 | 6 - 7 | 1052 | 123 | ||
| XM_005169229.4 | XP_005169286.1 | 7 - 8 | 1170 | 123 | ||
| XM_009305677.3 | XP_009303952.1 | 6 - 7 | 1055 | 123 | ||
| XM_009305679.3 | XP_009303954.1 | 5 - 6 | 639 | 123 | ||
| XM_009305680.3 | XP_009303955.1 | 5 - 6 | 635 | 123 | ||
| GenBank mRNA | BC059188.1 | - | 7 - 8 | 1122 | 123 | |
| BC070007.1 | - | 6 - 7 | 989 | 123 | ||
| KT020763.1 | - | 5 - 6 | 748 | 123 | ||
More Information
Set Membership: |
|

mef2ca
Genomic Map