Assay Details
| Gene Symbol: mef2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131312.2 | NP_571387.2 | 8 - 9 | 1292 | 61 | |
| XM_005169228.4 | XP_005169285.1 | 7 - 8 | 1222 | 61 | ||
| XM_005169232.4 | XP_005169289.1 | 7 - 8 | 1288 | 61 | ||
| GenBank mRNA | BC059188.1 | - | 8 - 9 | 1292 | 61 | |
| BC070007.1 | - | 7 - 8 | 1159 | 61 | ||
| KF932281.1 | - | 6 - 7 | 812 | 61 | ||
| KF932282.1 | - | 5 - 6 | 659 | 61 | ||
| U66569.1 | - | 5 - 6 | 815 | 61 | ||
More Information
Set Membership: |
|

mef2ca
Genomic Map