Assay Details
| Gene Symbol: mef2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131312.2 | NP_571387.2 | 9 - 10 | 1418 | 65 | |
| XM_005169228.4 | XP_005169285.1 | 8 - 9 | 1348 | 65 | ||
| XM_005169229.4 | XP_005169286.1 | 10 - 11 | 1490 | 65 | ||
| XM_005169232.4 | XP_005169289.1 | 8 - 9 | 1414 | 65 | ||
| XM_009305677.3 | XP_009303952.1 | 9 - 10 | 1375 | 65 | ||
| XM_009305679.3 | XP_009303954.1 | 8 - 9 | 959 | 65 | ||
| XM_009305680.3 | XP_009303955.1 | 8 - 9 | 955 | 65 | ||
| XM_009305681.3 | XP_009303956.1 | 9 - 10 | 1443 | 65 | ||
| GenBank mRNA | BC059188.1 | - | 9 - 10 | 1418 | 65 | |
| BC070007.1 | - | 8 - 9 | 1285 | 65 | ||
| KF932281.1 | - | 7 - 8 | 938 | 65 | ||
| KF932282.1 | - | 6 - 7 | 785 | 65 | ||
| U66569.1 | - | 6 - 7 | 941 | 65 | ||
More Information
Set Membership: |
|

mef2ca
Genomic Map