Assay Details
| Gene Symbol: mef2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131312.2 | NP_571387.2 | 10 - 11 | 1545 | 94 | |
| XM_005169228.4 | XP_005169285.1 | 9 - 10 | 1475 | 94 | ||
| XM_005169229.4 | XP_005169286.1 | 11 - 12 | 1617 | 94 | ||
| XM_005169232.4 | XP_005169289.1 | 9 - 10 | 1541 | 94 | ||
| XM_009305677.3 | XP_009303952.1 | 10 - 11 | 1502 | 94 | ||
| XM_009305679.3 | XP_009303954.1 | 9 - 10 | 1086 | 94 | ||
| XM_009305680.3 | XP_009303955.1 | 9 - 10 | 1082 | 94 | ||
| XM_009305681.3 | XP_009303956.1 | 10 - 11 | 1570 | 94 | ||
| GenBank mRNA | BC059188.1 | - | 10 - 11 | 1545 | 94 | |
| BC070007.1 | - | 9 - 10 | 1412 | 94 | ||
| KT020763.1 | - | 7 - 8 | 889 | 94 | ||
More Information
Set Membership: |
|

mef2ca
Genomic Map