Assay Details
| Gene Symbol: epha2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131415.1 | NP_571490.1 | 12 - 13 | 2173 | 67 | |
| GenBank mRNA | AJ005027.1 | - | 8 - 9 | 1187 | 67 | |
| BC065990.1 | - | 12 - 13 | 2173 | 67 | ||
More Information
Set Membership: |
|

epha2a
Genomic Map