Assay Details
| Gene Symbol: spon1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131515.1 | NP_571590.1 | 14 - 15 | 2144 | 77 | |
| GenBank mRNA | AB006086.1 | - | 14 - 15 | 2144 | 77 | |
| BC162586.1 | - | 14 - 15 | 2127 | 77 | ||
| KM655720.1 | - | 14 - 15 | 1992 | 77 | ||
More Information
Set Membership: |
|

spon1a
Genomic Map