Assay Details
| Gene Symbol: fabp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131605.2 | NP_571680.1 | 1 - 2 | 132 | 102 | |
| GenBank mRNA | AF237712.2 | - | 1 - 2 | 132 | 102 | |
| BC055621.1 | - | 1 - 2 | 135 | 102 | ||
| BC164937.1 | - | 1 - 2 | 105 | 102 | ||
More Information
Set Membership: |
|

fabp7a
Genomic Map