Assay Details
| Gene Symbol: tradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131607.2 | NP_571682.1 | 3 - 4 | 284 | 122 | |
| GenBank mRNA | BC083425.1 | - | 3 - 4 | 284 | 122 | |
| BC164053.1 | - | 2 - 3 | 203 | 122 | ||
More Information
Set Membership: |
|

tradd
Genomic Map