Assay Details
| Gene Symbol: pbx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131614.1 | NP_571689.1 | 6 - 7 | 1415 | 63 | |
| XM_017358962.2 | XP_017214451.1 | 6 - 7 | 1424 | 63 | ||
| GenBank mRNA | AJ245962.1 | - | 6 - 7 | 1415 | 63 | |
| BC066446.1 | - | 6 - 7 | 1378 | 63 | ||
More Information
Set Membership: |
|

pbx1a
Genomic Map