Assay Details
| Gene Symbol: atp1b3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131670.1 | NP_571745.1 | 4 - 5 | 603 | 128 | |
| GenBank mRNA | AF293369.1 | - | 4 - 5 | 603 | 128 | |
| BC076055.1 | - | 4 - 5 | 567 | 128 | ||
More Information
Set Membership: |
|

atp1b3b
Genomic Map