Assay Details
| Gene Symbol: nots | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131804.1 | NP_571879.1 | 801 | 90 | ||
| GenBank mRNA | AJ278269.1 | - | 801 | 90 | ||
| BC086835.1 | - | 966 | 90 | |||
| BC165335.1 | - | 916 | 90 | |||
More Information
Set Membership: |
|

nots
Genomic Map