Assay Details
| Gene Symbol: dmbx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152977.1 | NP_694509.1 | 3 - 4 | 433 | 61 | |
| NM_178163.2 | NP_835457.2 | 3 - 4 | 418 | 61 | ||
| GenBank mRNA | AF398525.1 | - | 3 - 4 | 433 | 61 | |
| AF398526.1 | - | 3 - 4 | 418 | 61 | ||
| AY071922.1 | - | 3 - 4 | 511 | 61 | ||
| BC134894.1 | - | 3 - 4 | 496 | 61 | ||
More Information
Set Membership: |
|

dmbx1a
Genomic Map