Assay Details
| Gene Symbol: esr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180966.2 | NP_851297.1 | 2 - 3 | 536 | 129 | |
| GenBank mRNA | AF349414.1 | - | 2 - 3 | 519 | 129 | |
| AF516874.1 | - | 3 - 4 | 692 | 129 | ||
| AJ414567.1 | - | 2 - 3 | 536 | 129 | ||
| AY307098.1 | - | 1 - 2 | 398 | 129 | ||
| BC044349.1 | - | 2 - 3 | 536 | 129 | ||
| BC165404.1 | - | 1 - 2 | 432 | 129 | ||
More Information
Set Membership: |
|

esr2a
Genomic Map