Assay Details
| Gene Symbol: esr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180966.2 | NP_851297.1 | 4 - 5 | 826 | 74 | |
| GenBank mRNA | AJ414567.1 | - | 4 - 5 | 826 | 74 | |
| AY307098.1 | - | 3 - 4 | 688 | 74 | ||
| BC044349.1 | - | 4 - 5 | 826 | 74 | ||
| BC165404.1 | - | 3 - 4 | 722 | 74 | ||
More Information
Set Membership: |
|

esr2a
Genomic Map