Assay Details
| Gene Symbol: esr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180966.2 | NP_851297.1 | 5 - 6 | 1145 | 94 | |
| GenBank mRNA | AF349414.1 | - | 5 - 6 | 1128 | 94 | |
| AF516874.1 | - | 6 - 7 | 1301 | 94 | ||
| AJ414567.1 | - | 5 - 6 | 1145 | 94 | ||
| AY307098.1 | - | 4 - 5 | 1007 | 94 | ||
| BC044349.1 | - | 5 - 6 | 1145 | 94 | ||
| BC165404.1 | - | 4 - 5 | 1041 | 94 | ||
More Information
Set Membership: |
|

esr2a
Genomic Map