Assay Details
| Gene Symbol: esr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180966.2 | NP_851297.1 | 8 - 9 | 1600 | 61 | |
| GenBank mRNA | AF516874.1 | - | 9 - 10 | 1756 | 61 | |
| AJ414567.1 | - | 8 - 9 | 1600 | 61 | ||
| AY307098.1 | - | 7 - 8 | 1462 | 61 | ||
| BC044349.1 | - | 8 - 9 | 1600 | 61 | ||
| BC165404.1 | - | 7 - 8 | 1496 | 61 | ||
More Information
Set Membership: |
|

esr2a
Genomic Map