Assay Details
| Gene Symbol: anxa2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181761.2 | NP_861426.1 | 12 - 13 | 1005 | 84 | |
| GenBank mRNA | AY178796.1 | - | 12 - 13 | 991 | 84 | |
| BC056699.1 | - | 12 - 13 | 1005 | 84 | ||
| BC062531.1 | - | 12 - 13 | 997 | 84 | ||
More Information
Set Membership: |
|

anxa2a
Genomic Map