Assay Details
| Gene Symbol: slc34a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182877.2 | NP_878297.1 | 1 - 2 | 56 | 147 | |
| XM_021469757.1 | XP_021325432.1 | 2 - 3 | 506 | 147 | ||
| GenBank mRNA | AF297180.2 | - | 1 - 2 | 57 | 147 | |
| BC054940.1 | - | 2 - 3 | 75 | 147 | ||
More Information
Set Membership: |
|

slc34a2b
Genomic Map