Assay Details
| Gene Symbol: slc34a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182877.2 | NP_878297.1 | 4 - 5 | 479 | 58 | |
| XM_021469757.1 | XP_021325432.1 | 5 - 6 | 929 | 58 | ||
| GenBank mRNA | AF297180.2 | - | 4 - 5 | 480 | 58 | |
More Information
Set Membership: |
|

slc34a2b
Genomic Map