Assay Details
| Gene Symbol: slc34a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182877.2 | NP_878297.1 | 8 - 9 | 999 | 72 | |
| XM_021469757.1 | XP_021325432.1 | 9 - 10 | 1449 | 72 | ||
| GenBank mRNA | AF297180.2 | - | 8 - 9 | 1000 | 72 | |
| BC054940.1 | - | 9 - 10 | 1018 | 72 | ||
More Information
Set Membership: |
|

slc34a2b
Genomic Map