Assay Details
| Gene Symbol: aup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199690.2 | NP_955984.2 | 5 - 6 | 704 | 63 | |
| XM_005173083.4 | XP_005173140.1 | 5 - 6 | 746 | 63 | ||
| XM_009307693.3 | XP_009305968.1 | 5 - 6 | 749 | 63 | ||
| GenBank mRNA | BC059643.1 | - | 5 - 6 | 702 | 63 | |
| BC065447.1 | - | 5 - 6 | 704 | 63 | ||
More Information
Set Membership: |
|

aup1
Genomic Map