Assay Details
| Gene Symbol: st13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199769.2 | NP_956063.1 | 11 - 12 | 1009 | 180 | |
| GenBank mRNA | EB881211.1 | - | 2 - 3 | 155 | 180 | |
More Information
Set Membership: |
|

st13
Genomic Map