Assay Details
| Gene Symbol: st13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199769.2 | NP_956063.1 | 12 - 13 | 1091 | 85 | |
| GenBank mRNA | EB881211.1 | - | 3 - 4 | 237 | 85 | |
More Information
Set Membership: |
|

st13
Genomic Map