Assay Details
| Gene Symbol: st13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199769.2 | NP_956063.1 | 8 - 9 | 718 | 63 | |
| GenBank mRNA | BC063322.1 | - | 8 - 9 | 702 | 63 | |
| BC067180.1 | - | 8 - 9 | 703 | 63 | ||
| DR728308.1 | - | 6 - 7 | 471 | 63 | ||
More Information
Set Membership: |
|

st13
Genomic Map