Assay Details
| Gene Symbol: st13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199769.2 | NP_956063.1 | 9 - 10 | 829 | 65 | |
| GenBank mRNA | BC063322.1 | - | 9 - 10 | 813 | 65 | |
| BC067180.1 | - | 9 - 10 | 814 | 65 | ||
| DR728308.1 | - | 7 - 8 | 582 | 65 | ||
More Information
Set Membership: |
|

st13
Genomic Map