Assay Details
| Gene Symbol: ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199785.1 | NP_956079.1 | 9 - 10 | 1934 | 63 | |
| GenBank mRNA | BC054895.1 | - | 9 - 10 | 1934 | 63 | |
More Information
Set Membership: |
|

ercc4
Genomic Map