Assay Details
| Gene Symbol: iars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199896.2 | NP_956190.2 | 26 - 27 | 2844 | 71 | |
| GenBank mRNA | BC056826.1 | - | 26 - 27 | 2838 | 71 | |
| DR716413.1 | - | 3 - 4 | 378 | 71 | ||
More Information
Set Membership: |
|

iars1
Genomic Map