Assay Details
| Gene Symbol: iars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199896.2 | NP_956190.2 | 29 - 30 | 3229 | 70 | |
| GenBank mRNA | BC056826.1 | - | 29 - 30 | 3223 | 70 | |
| EH601215.1 | - | 5 - 6 | 728 | 70 | ||
More Information
Set Membership: |
|

iars1
Genomic Map