Assay Details
| Gene Symbol: iars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199896.2 | NP_956190.2 | 30 - 31 | 3362 | 84 | |
| GenBank mRNA | BC056826.1 | - | 30 - 31 | 3356 | 84 | |
| EH601215.1 | - | 4 - 5 | 595 | 84 | ||
More Information
Set Membership: |
|

iars1
Genomic Map