Assay Details
| Gene Symbol: iars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199896.2 | NP_956190.2 | 24 - 25 | 2580 | 64 | |
| GenBank mRNA | BC056826.1 | - | 24 - 25 | 2574 | 64 | |
| DR716413.1 | - | 1 - 2 | 114 | 64 | ||
More Information
Set Membership: |
|

iars1
Genomic Map