Assay Details
| Gene Symbol: strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199911.2 | NP_956205.2 | 11 - 12 | 1289 | 67 | |
| XM_021469637.1 | XP_021325312.1 | 12 - 13 | 1328 | 67 | ||
| XM_021469639.1 | XP_021325314.1 | 12 - 13 | 1328 | 67 | ||
| GenBank mRNA | BC047830.1 | - | 10 - 11 | 936 | 67 | |
More Information
Set Membership: |
|

strada
Genomic Map