Assay Details
| Gene Symbol: irf2bp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199981.1 | NP_956275.1 | 1 - 2 | 975 | 75 | |
| GenBank mRNA | BC046025.1 | - | 1 - 2 | 975 | 75 | |
| BC065975.1 | - | 1 - 2 | 885 | 75 | ||
More Information
Set Membership: |
|

irf2bp2a
Genomic Map