Assay Details
| Gene Symbol: ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200093.1 | NP_956387.1 | 3 - 4 | 406 | 123 | |
| GenBank mRNA | BC059514.1 | - | 3 - 4 | 406 | 123 | |
| BC065474.1 | - | 3 - 4 | 379 | 123 | ||
| BC068376.1 | - | 3 - 4 | 401 | 123 | ||
More Information
Set Membership: |
|

ormdl1
Genomic Map