Assay Details
| Gene Symbol: eif1ad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200468.1 | NP_956762.1 | 3 - 4 | 268 | 131 | |
| GenBank mRNA | BC055173.1 | - | 3 - 4 | 268 | 131 | |
More Information
Set Membership: |
|

eif1ad
Genomic Map