Assay Details
| Gene Symbol: zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200517.2 | NP_956811.1 | 3 - 4 | 306 | 79 | |
| XM_005164201.4 | XP_005164258.1 | 4 - 5 | 482 | 79 | ||
| XM_005164202.4 | XP_005164259.1 | 4 - 5 | 484 | 79 | ||
| XM_009299701.3 | XP_009297976.1 | 4 - 5 | 489 | 79 | ||
| XM_021469821.1 | XP_021325496.1 | 4 - 5 | 483 | 79 | ||
| GenBank mRNA | BC055589.1 | - | 4 - 5 | 430 | 79 | |
| BC065896.1 | - | 3 - 4 | 306 | 79 | ||
More Information
Set Membership: |
|

zfand2a
Genomic Map