Assay Details
| Gene Symbol: polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200566.1 | NP_956860.1 | 7 - 8 | 814 | 75 | |
| GenBank mRNA | AY648731.1 | - | 7 - 8 | 75 | ||
| BC056581.1 | - | 7 - 8 | 814 | 75 | ||
More Information
Set Membership: |
|

polr1c
Genomic Map