Assay Details
| Gene Symbol: rgs9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200830.2 | NP_957124.2 | 3 - 4 | 272 | 142 | |
| XM_021480167.1 | XP_021335842.1 | 3 - 4 | 398 | 142 | ||
| XM_021480168.1 | XP_021335843.1 | 3 - 4 | 396 | 142 | ||
| GenBank mRNA | BC060915.1 | - | 3 - 4 | 273 | 142 | |
| BC152204.1 | - | 3 - 4 | 326 | 142 | ||
More Information
Set Membership: |
|

rgs9b
Genomic Map